Assay Details
| Gene Symbol: CXCL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000609.6 | NP_000600.1 | 3 - 4 | 349 | 71 | |
| GenBank mRNA | L36033.1 | - | 3 - 4 | 336 | 71 | |
| U16752.1 | - | 3 - 4 | 337 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001212.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
neuron migration
positive regulation of endothelial cell proliferation
cellular calcium ion homeostasis
chemotaxis
immune response
cell adhesion
signal transduction
G-protein coupled receptor signaling pathway
axon guidance
blood circulation
regulation of actin polymerization or depolymerization
adult locomotory behavior
response to radiation
response to heat
response to mechanical stimulus
response to virus
telencephalon cell migration
organ regeneration
positive regulation of dopamine secretion
response to peptide hormone
positive regulation of cell adhesion
positive regulation of axon extension involved in axon guidance
positive chemotaxis
induction of positive chemotaxis
cell chemotaxis
chemokine-mediated signaling pathway
positive regulation of monocyte chemotaxis
positive regulation of calcium ion import
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of leukocyte tethering or rolling
negative regulation of leukocyte apoptotic process
positive regulation of T cell migration
receptor binding
chemokine activity
growth factor activity
chemoattractant activity
chemokine receptor binding
CXCR chemokine receptor binding
response to hypoxia
neuron migration
positive regulation of endothelial cell proliferation
cellular calcium ion homeostasis
chemotaxis
immune response
cell adhesion
signal transduction
G-protein coupled receptor signaling pathway
axon guidance
blood circulation
regulation of actin polymerization or depolymerization
adult locomotory behavior
response to radiation
response to heat
response to mechanical stimulus
response to virus
telencephalon cell migration
organ regeneration
positive regulation of dopamine secretion
response to peptide hormone
positive regulation of cell adhesion
positive regulation of axon extension involved in axon guidance
positive chemotaxis
induction of positive chemotaxis
cell chemotaxis
chemokine-mediated signaling pathway
positive regulation of monocyte chemotaxis
positive regulation of calcium ion import
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of leukocyte tethering or rolling
negative regulation of leukocyte apoptotic process
positive regulation of T cell migration
receptor binding
chemokine activity
growth factor activity
chemoattractant activity
chemokine receptor binding
CXCR chemokine receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |

CXCL12
Genomic Map