| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019649.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019649.hg.1 |
ENST00000368482.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019649.hg.1 |
ENST00000479937.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019649.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019649.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019649.hg.1 |
TC01001277.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100173329.hg.1 |
TC0100010082.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368480.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000368482.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000462871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000468714.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
ENST00000479937.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_001098475.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010082.hg.1 |
NM_182499.3 |
Refseq |