Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320263.1 | NP_001307192.1 | 4 - 5 | 895 | 87 | |
| NM_001320264.1 | NP_001307193.1 | 2 - 3 | 734 | 87 | ||
| NM_012095.5 | NP_036227.1 | 2 - 3 | 608 | 87 | ||
| NM_207012.3 | NP_996895.1 | 3 - 4 | 734 | 87 | ||
| NR_135191.1 | 2 - 3 | 608 | 87 | |||
| GenBank mRNA | AF092092.1 | - | 1 - 2 | 274 | 87 | |
| AK026983.1 | - | 2 - 3 | 343 | 87 | ||
| AK315597.1 | - | 2 - 3 | 317 | 87 | ||
| BC026232.1 | - | 3 - 4 | 493 | 4807259 | 87 | |
| BC067127.1 | - | 2 - 3 | 704 | 6169822 | 87 | |
| BU172918.1 | - | 2 - 3 | 572 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001423.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10018067.hg.1 | ENST00000355264.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018067.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018067.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018067.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018067.hg.1 | TC10001423.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018068.hg.1 | ENST00000355264.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018068.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018068.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018068.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018068.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018068.hg.1 | TC10001423.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018074.hg.1 | ENST00000355264.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018074.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018074.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018074.hg.1 | TC10001423.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018069.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018069.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018069.hg.1 | TC10001423.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018073.hg.1 | ENST00000355264.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018073.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018073.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018073.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018073.hg.1 | TC10001423.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018065.hg.1 | ENST00000355264.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018065.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018065.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018065.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018065.hg.1 | TC10001423.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000480373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000480373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000480373.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | ENST00000598318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000480373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000480373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000480373.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | ENST00000598318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011050.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175426.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175431.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175431.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175431.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175434.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175427.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | ENST00000487653.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175428.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175430.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175430.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175430.hg.1 | ENST00000372745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175430.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175430.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | ENST00000355264.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | NM_012095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | NM_207012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000175435.hg.1 | TC1000011050.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 216588 | - | - |

AP3M1
Genomic Map