| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018059.hg.1 |
ENST00000355264.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018059.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018059.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018059.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10018059.hg.1 |
TC10001423.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000480373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000480373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000480373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000487653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000487653.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
ENST00000598318.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000480373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000480373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000480373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000487653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000487653.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
ENST00000598318.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011050.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
ENST00000355264.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
ENST00000372745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
NM_012095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
NM_207012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000175420.hg.1 |
TC1000011050.hg.1 |
Clariom D TC |