Assay Details
| Gene Symbol: COL9A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001851.4 | NP_001842.3 | 12 - 13 | 1220 | 144 | |
| NM_078485.3 | NP_511040.2 | 6 - 7 | 488 | 144 | ||
| XM_011535429.2 | XP_011533731.1 | 12 - 13 | 1220 | 144 | ||
| XM_011535430.2 | XP_011533732.1 | 6 - 7 | 486 | 144 | ||
| XM_017010246.1 | XP_016865735.1 | 9 - 10 | 577 | 144 | ||
| XM_017010247.1 | XP_016865736.1 | 3 - 4 | 433 | 144 | ||
| GenBank mRNA | AK125738.1 | - | 3 - 4 | 207 | 144 | |
| BC008620.1 | - | 6 - 7 | 415 | 144 | ||
| BC063646.1 | - | 6 - 7 | 462 | 4914907 | 144 | |
| CA392556.1 | - | 6 - 7 | 488 | 144 | ||
| X54412.1 | - | 12 - 13 | 1192 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK125738.1, XM_017010247.1 transcripts.
Set Membership: |
|

COL9A1
Genomic Map