Assay Details
| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040515.1 | 1 - 2 | 283 | 64 | ||
| XM_006723226.2 | XP_006723289.2 | 1 - 2 | 275 | 64 | ||
| GenBank mRNA | AK057862.1 | - | 1 - 2 | 234 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

NFKBIB
Genomic Map