Assay Details
| Gene Symbol: KLK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102274.1 | 3 - 4 | 394 | 52 | ||
| XM_017026943.1 | XP_016882432.1 | 4 - 5 | 1208 | 52 | ||
| GenBank mRNA | AY373373.1 | - | 3 | 290 | ||
| BC144045.1 | - | 3 - 4 | 384 | 52 | ||
| X75363.1 | - | 1 | 287 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000598673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | TC1900011224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194332.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000598673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011224.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NR_102274.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | TC1900011224.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000326856.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000596531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000596931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000598239.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000601680.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | ENST00000602114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NM_001277081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NM_001277082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NM_017509.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194335.hg.1 | NR_102274.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 159271 | - | - |

KLK15
Genomic Map