Assay Details
| Gene Symbol: STARD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139171.1 | NP_631910.1 | 1 - 2 | 82 | 155 | |
| XM_011525821.1 | XP_011524123.1 | 3 - 4 | 269 | 155 | ||
| GenBank mRNA | AF480305.1 | - | 1 - 2 | 82 | 155 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000523.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18005436.hg.1 | ENST00000307844.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005436.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005436.hg.1 | TC18000523.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005437.hg.1 | ENST00000307844.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005437.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005437.hg.1 | TC18000523.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008734.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | TC1800008734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159893.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | NM_139171.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | TC1800008734.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000307844.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000577499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000581310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | ENST00000584040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159891.hg.1 | NM_139171.1 | Refseq |

STARD6
Genomic Map