Assay Details
| Gene Symbol: PPP2R2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271900.1 | NP_001258829.1 | 2 - 3 | 347 | 91 | |
| GenBank mRNA | AK294659.1 | - | 2 - 3 | 256 | 91 | |
More Information
Set Membership: |
|

PPP2R2B
Genomic Map