Assay Details
| Gene Symbol: NDFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030571.3 | NP_085048.1 | 1 - 2 | 284 | 134 | |
| GenBank mRNA | AB097010.1 | - | 1 - 2 | 220 | 134 | |
| AB097037.1 | - | 1 - 2 | 220 | 134 | ||
| AI929639.1 | - | 1 - 2 | 284 | 134 | ||
| AK075495.1 | - | 2 - 3 | 204 | 134 | ||
| AK315481.1 | - | 1 - 2 | 229 | 134 | ||
| AY192728.1 | - | 1 - 2 | 240 | 134 | ||
| BC004317.1 | - | 1 - 2 | 172 | 3628100 | 134 | |
| DA236494.1 | - | 1 - 2 | 284 | 134 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000777.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB097010.1,AB097037.1,AK075495.1,AK315481.1,AY192728.1,BC004317.1,NM_030571.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of myeloid leukocyte differentiation
negative regulation of type 2 immune response
cellular iron ion homeostasis
vacuolar transport
signal transduction
negative regulation of gene expression
metal ion transport
positive regulation of protein ubiquitination
negative regulation of transporter activity
negative regulation of interleukin-4 production
negative regulation of T cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
regulation of lymphocyte differentiation
positive regulation of protein catabolic process
negative regulation of isotype switching to IgE isotypes
regulation of isotype switching to IgG isotypes
negative regulation of inflammatory response
negative regulation of protein transport
signal transducer activity
protein binding
regulation of myeloid leukocyte differentiation
negative regulation of type 2 immune response
cellular iron ion homeostasis
vacuolar transport
signal transduction
negative regulation of gene expression
metal ion transport
positive regulation of protein ubiquitination
negative regulation of transporter activity
negative regulation of interleukin-4 production
negative regulation of T cell proliferation
positive regulation of I-kappaB kinase/NF-kappaB signaling
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
regulation of lymphocyte differentiation
positive regulation of protein catabolic process
negative regulation of isotype switching to IgE isotypes
regulation of isotype switching to IgG isotypes
negative regulation of inflammatory response
negative regulation of protein transport
signal transducer activity
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000505614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000505614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000505614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011081.hg.1 | ENST00000253814.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011081.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011081.hg.1 | TC05000777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011083.hg.1 | ENST00000253814.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011083.hg.1 | ENST00000313129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011083.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011083.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011083.hg.1 | TC05000777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011085.hg.1 | ENST00000253814.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011085.hg.1 | ENST00000313129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011085.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011085.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011085.hg.1 | TC05000777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011084.hg.1 | ENST00000253814.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011084.hg.1 | ENST00000313129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011084.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011084.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011084.hg.1 | TC05000777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010064.hg.1 | ENST00000543144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011087.hg.1 | ENST00000253814.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011087.hg.1 | ENST00000313129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011087.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011087.hg.1 | TC05000777.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200010064.hg.1 | ENST00000543144.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162199.hg.1 | TC0500008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162203.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162203.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162203.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162203.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162203.hg.1 | TC0500008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000503388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000505614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000505614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000505614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008921.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162201.hg.1 | TC0500008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162200.hg.1 | TC0500008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | ENST00000509436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162198.hg.1 | TC0500008921.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500162197.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162197.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162197.hg.1 | ENST00000253814.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500162197.hg.1 | NM_030571.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500162197.hg.1 | TC0500008921.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 119905 | - | - |

NDFIP1
Genomic Map