| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011092.hg.1 |
ENST00000253814.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011092.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05011092.hg.1 |
TC05000777.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000503388.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
ENST00000509436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008921.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
ENST00000253814.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
ENST00000505614.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
NM_030571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500162206.hg.1 |
TC0500008921.hg.1 |
Clariom D TC |