Assay Details
| Gene Symbol: P2RY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002564.3 | NP_002555.3 | 3 | 6840 | 85 | |
| NM_176071.2 | NP_788085.2 | 3 | 6905 | 85 | ||
| NM_176072.2 | NP_788086.2 | 3 | 6974 | 85 | ||
| GenBank mRNA | AK094727.1 | - | 1 | 1691 | 85 | |
| BX641058.1 | - | 1 | 574 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
cellular ion homeostasis
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
aging
positive regulation of neuron projection development
positive regulation of smooth muscle cell migration
sensory perception of pain
negative regulation of ossification
positive regulation of prostaglandin secretion
positive regulation of chemokine production
G-protein coupled purinergic nucleotide receptor signaling pathway
wound healing
regulation of vasodilation
positive regulation of penile erection
positive regulation of mucus secretion
response to growth factor
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to purine-containing compound
receptor activity
ATP binding
G-protein coupled purinergic nucleotide receptor activity
UTP-activated nucleotide receptor activity
in utero embryonic development
cellular ion homeostasis
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
aging
positive regulation of neuron projection development
positive regulation of smooth muscle cell migration
sensory perception of pain
negative regulation of ossification
positive regulation of prostaglandin secretion
positive regulation of chemokine production
G-protein coupled purinergic nucleotide receptor signaling pathway
wound healing
regulation of vasodilation
positive regulation of penile erection
positive regulation of mucus secretion
response to growth factor
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to purine-containing compound
receptor activity
ATP binding
G-protein coupled purinergic nucleotide receptor activity
UTP-activated nucleotide receptor activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000311131.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000393597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008378.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161930.hg.1 | TC1100008378.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161929.hg.1 | TC1100008378.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | ENST00000565433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | NM_002564.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | NM_176071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | NM_176072.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100161928.hg.1 | TC1100008378.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 173987 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 232334 | - | - |

P2RY2
Genomic Map