Assay Details
| Gene Symbol: PLXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282736.1 | NP_001269665.1 | 2 - 3 | 967 | 98 | |
| GenBank mRNA | BC012885.1 | - | 2 - 3 | 573 | 3865073 | 98 |
More Information
Set Membership: |
|

PLXDC2
Genomic Map