| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
ENST00000371426.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
ENST00000371430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
ENST00000394100.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
ENST00000536233.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019356.hg.1 |
TC10001538.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000460752.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000462771.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000482719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
ENST00000483229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011454.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177761.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177761.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177761.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177761.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177761.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
ENST00000359204.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
ENST00000472153.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
NM_145246.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177763.hg.1 |
TC1000011454.hg.1 |
Clariom D TC |