Assay Details
| Gene Symbol: KAZALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319303.1 | NP_001306232.1 | 3 - 4 | 1657 | 118 | |
| NM_030929.4 | NP_112191.2 | 3 - 4 | 1602 | 118 | ||
| NR_135067.1 | 2 - 3 | 1040 | 118 | |||
| XM_017016714.1 | XP_016872203.1 | 2 - 3 | 681 | 118 | ||
| XM_017016715.1 | XP_016872204.1 | 2 - 3 | 681 | 118 | ||
| GenBank mRNA | AK172862.1 | - | 3 - 4 | 853 | 118 | |
| AK172864.1 | - | 3 - 4 | 1656 | 118 | ||
| AK225949.1 | - | 3 - 4 | 854 | 118 | ||
| AY359087.1 | - | 3 - 4 | 858 | 118 | ||
| BC007758.2 | - | 3 - 4 | 1003 | 4122366 | 118 | |
| BI766345.1 | - | 2 - 3 | 313 | 118 | ||
| CX871488.1 | - | 2 - 3 | 331 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000732.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10008643.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008643.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008643.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008631.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008631.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008631.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008631.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008642.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008642.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008642.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008630.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008630.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008630.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008630.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008634.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008634.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008634.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008634.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008632.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008632.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008632.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008632.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008637.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008637.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008637.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008637.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008639.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008639.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008639.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008639.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008641.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008641.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008641.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008629.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008629.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008629.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008635.hg.1 | ENST00000313664.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008635.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008635.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008635.hg.1 | TC10000732.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012493.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186186.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186188.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186188.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186188.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186188.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186188.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186192.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186194.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186187.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186184.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186184.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186184.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | ENST00000470106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186190.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186185.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186185.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186185.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186185.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186185.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186189.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186189.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186189.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186189.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186189.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | ENST00000370200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | ENST00000477979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | NM_030929.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186193.hg.1 | TC1000012493.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 171900 | - | - |

KAZALD1
Genomic Map