Assay Details
| Gene Symbol: PDLIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011516.2 | NP_001011516.1 | 3 - 4 | 432 | 138 | |
| XM_006714070.3 | XP_006714133.1 | 3 - 4 | 405 | 138 | ||
| GenBank mRNA | AK291898.1 | - | 3 - 4 | 357 | 138 | |
| BC017902.1 | - | 3 - 4 | 375 | 138 | ||
| DA895978.1 | - | 3 - 4 | 432 | 138 | ||
More Information
Set Membership: |
|

PDLIM5
Genomic Map