| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06029098.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06029098.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06029098.hg.1 |
TC06002318.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06029097.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06029097.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06029097.hg.1 |
TC06002318.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013942.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196074.hg.1 |
TC0600013942.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
ENST00000366822.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
ENST00000495520.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
NR_026773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196072.hg.1 |
TC0600013942.hg.1 |
Clariom D TC |