| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023776.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023776.hg.1 |
ENST00000536477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023776.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023776.hg.1 |
TC06001940.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600186427.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186427.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186427.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600186427.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600186427.hg.1 |
TC0600012542.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
ENST00000435041.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012542.hg.1 |
NM_016021.2 |
Refseq |