Assay Details
| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020429.1 | XP_016875918.1 | 2 - 3 | 580 | 113 | |
| XM_017020430.1 | XP_016875919.1 | 2 - 3 | 454 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LRRC63
Genomic Map