Assay Details
| Gene Symbol: LINC01187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108022.1 | 4 - 5 | 717 | 97 | ||
| GenBank mRNA | AK055811.1 | - | 3 - 4 | 1628 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box I1 | FOXI1 |
FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3 |
-81.9kb | Anti-sense | Chr. 5 - 170105897 - 170109725 on NCBI Build GRCh38 |
| Downstream | chromosome 5 open reading frame 58 | C5orf58 | 38.5kb | Anti-sense | Chr. 5 - 170231730 - 170252569 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01187
Genomic Map