| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
ENST00000600282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024401.hg.1 |
ENST00000330123.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024401.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024401.hg.1 |
TC19001796.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000596290.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000597013.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
ENST00000600282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012022.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207671.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207671.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207671.hg.1 |
TC1900012022.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207671.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207671.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
NM_178523.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
TC1900012022.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
ENST00000330123.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
ENST00000600228.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207672.hg.1 |
NM_178523.4 |
Refseq |