| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024934.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024934.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024934.hg.1 |
TC06002004.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_001164313.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600014332.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202447.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202447.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202447.hg.1 |
ENST00000230122.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202447.hg.1 |
NM_014797.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202447.hg.1 |
TC0600014332.hg.1 |
Clariom D TC |