| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15005809.hg.1 |
ENST00000261921.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15005809.hg.1 |
ENST00000395162.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15005809.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15005809.hg.1 |
TC15000670.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500155647.hg.1 |
TC1500007829.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000261921.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000562548.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566011.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000566530.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
ENST00000567675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007829.hg.1 |
NM_005576.3 |
Refseq |