Assay Details
| Gene Symbol: RGS7BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029875.2 | NP_001025046.1 | 4 - 5 | 1281 | 86 | |
| NM_001271890.1 | NP_001258819.1 | 2 - 3 | 395 | 86 | ||
| NR_073511.1 | 2 - 3 | 395 | 86 | |||
| GenBank mRNA | AK294808.1 | - | 3 - 4 | 386 | 86 | |
| AK296443.1 | - | 4 - 5 | 929 | 86 | ||
| AK296494.1 | - | 2 - 3 | 363 | 86 | ||
| BC140707.1 | - | 4 - 5 | 627 | 9021575 | 86 | |
| BX640900.1 | - | 4 - 5 | 519 | 86 | ||
| DA417118.1 | - | 2 - 3 | 363 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151174.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151174.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151174.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151174.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151174.hg.1 | TC0500007586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151173.hg.1 | TC0500007586.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000334025.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000505263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | ENST00000508162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001029875.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NM_001271891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007586.hg.1 | NR_073511.1 | Refseq |

RGS7BP
Genomic Map