Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184891.1 | NP_001171820.1 | 1 - 2 | 105 | 72 | |
| NM_001184892.1 | NP_001171821.1 | 1 - 2 | 105 | 72 | ||
| NM_201398.2 | NP_958800.1 | 1 - 2 | 105 | 72 | ||
| GenBank mRNA | AK025979.1 | - | 1 - 2 | 33 | 72 | |
| AK308243.1 | - | 1 - 2 | 39 | 72 | ||
| BC011378.1 | - | 1 - 2 | 34 | 3855223 | 72 | |
| BC032323.1 | - | 1 - 2 | 40 | 5242727 | 72 | |
| BI919061.1 | - | 1 - 2 | 87 | 72 | ||
| DC371856.1 | - | 1 - 2 | 106 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001282.hg
Additional Information:
This assay is in the 5'UTR region of the BC011378.1,BC032323.1,NM_001184891.1,NM_001184892.1,NM_201398.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 209436 | - | - |

FLAD1
Genomic Map