Assay Details
| Gene Symbol: HGFAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297439.1 | NP_001284368.1 | 8 - 9 | 1083 | 167 | |
| NM_001528.3 | NP_001519.1 | 8 - 9 | 1083 | 167 | ||
| GenBank mRNA | AK314099.1 | - | 8 - 9 | 1041 | 167 | |
| BC112190.1 | - | 8 - 9 | 1083 | 8327658 | 167 | |
| BC112192.1 | - | 8 - 9 | 1095 | 8327660 | 167 | |
| D14012.1 | - | 8 - 9 | 1017 | 167 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000047.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04001303.hg.1 | ENST00000382774.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001303.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001303.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001303.hg.1 | TC04000047.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001307.hg.1 | ENST00000382774.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001307.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001307.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04001307.hg.1 | TC04000047.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000506132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006602.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | ENST00000382774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | ENST00000509689.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | ENST00000511533.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | NM_001297439.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | NM_001528.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400145779.hg.1 | TC0400006602.hg.1 | Clariom D TC |

HGFAC
Genomic Map