| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005901.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005901.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005901.hg.1 |
TC02000393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005903.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005903.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005903.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02005903.hg.1 |
TC02000393.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007872.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
ENST00000462772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154353.hg.1 |
TC0200007872.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200154351.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154351.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154351.hg.1 |
ENST00000272342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154351.hg.1 |
NM_019002.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154351.hg.1 |
TC0200007872.hg.1 |
Clariom D TC |