Assay Details
| Gene Symbol: ATP6V1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001693.3 | NP_001684.2 | 10 - 11 | 1284 | 95 | |
| GenBank mRNA | AK294163.1 | - | 11 - 12 | 1044 | 95 | |
| AK312372.1 | - | 10 - 11 | 1091 | 95 | ||
| BC003100.1 | - | 10 - 11 | 1086 | 3502278 | 95 | |
| BC007309.1 | - | 10 - 11 | 1041 | 95 | ||
| BC030640.2 | - | 10 - 11 | 1096 | 4821062 | 95 | |
| L35249.1 | - | 10 - 11 | 1073 | 95 | ||
| X62949.1 | - | 10 - 11 | 1095 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000151.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
ATP metabolic process
phagosome acidification
protein binding
ATP binding
hydrogen ion transmembrane transporter activity
proton-transporting ATPase activity, rotational mechanism
insulin receptor signaling pathway
ATP hydrolysis coupled proton transport
proton transport
regulation of macroautophagy
transferrin transport
ion transmembrane transport
ATP metabolic process
phagosome acidification
protein binding
ATP binding
hydrogen ion transmembrane transporter activity
proton-transporting ATPase activity, rotational mechanism
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08001560.hg.1 | ENST00000276390.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001560.hg.1 | ENST00000542368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001560.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001560.hg.1 | TC08000151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800147001.hg.1 | TC0800006933.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000276390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000519667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000520830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000521442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | ENST00000523482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006933.hg.1 | NM_001693.3 | Refseq |

ATP6V1B2
Genomic Map