Assay Details
| Gene Symbol: RCOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015156.3 | NP_055971.2 | 9 - 10 | 1356 | 103 | |
| GenBank mRNA | AF155595.1 | - | 9 - 10 | 1119 | 103 | |
| D31888.1 | - | 9 - 10 | 861 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000784.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
blood coagulation
viral process
negative regulation of transcription, DNA-templated
histone H4 deacetylation
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
blood coagulation
viral process
negative regulation of transcription, DNA-templated
histone H4 deacetylation
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14008496.hg.1 | ENST00000262241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008496.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008496.hg.1 | TC14000784.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000558495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000558495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000558495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000558495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000558495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000558495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | ENST00000560472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008355.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | ENST00000262241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | ENST00000559597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | NM_015156.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159632.hg.1 | TC1400008355.hg.1 | Clariom D TC |

RCOR1
Genomic Map