| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18001385.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18001385.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18001385.hg.1 |
TC18000124.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1800147976.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800147976.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800147976.hg.1 |
TC1800007010.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800147976.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800147976.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000257192.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
ENST00000462981.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007010.hg.1 |
NM_001942.3 |
Refseq |