Assay Details
| Gene Symbol: DSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001942.3 | NP_001933.2 | 6 - 7 | 903 | 125 | |
| GenBank mRNA | AF097935.1 | - | 6 - 7 | 904 | 125 | |
| X56654.1 | - | 6 - 7 | 768 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000124.hg.1
Set Membership: |
|

DSG1
Genomic Map