Assay Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC018772.1 | - | 9 - 10 | 2305 | 4869114 | 96 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564466.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000564526.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000566909.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | ENST00000569488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011075.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | NM_017740.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | TC1600011075.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | ENST00000313732.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | ENST00000344861.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | NM_001145548.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600186079.hg.1 | NM_017740.2 | Refseq |

ZDHHC7
Genomic Map