Assay Details
| Gene Symbol: EHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289413.1 | NP_001276342.1 | 21 - 22 | 3071 | 103 | |
| NM_001318833.1 | NP_001305762.1 | 23 - 24 | 3278 | 103 | ||
| NM_006709.4 | NP_006700.3 | 23 - 24 | 3028 | 103 | ||
| NM_025256.6 | NP_079532.5 | 22 - 23 | 2926 | 103 | ||
| XM_005248824.3 | XP_005248881.1 | 22 - 23 | 3169 | 103 | ||
| XM_006714974.1 | XP_006715037.1 | 22 - 23 | 3194 | 103 | ||
| XM_006714975.1 | XP_006715038.1 | 21 - 22 | 3092 | 103 | ||
| XM_006714976.1 | XP_006715039.1 | 23 - 24 | 3032 | 103 | ||
| XM_017010202.1 | XP_016865691.1 | 22 - 23 | 2930 | 103 | ||
| GenBank mRNA | AB209433.2 | - | 22 - 23 | 3111 | 103 | |
| AK092866.1 | - | 6 - 7 | 1943 | 103 | ||
| BC002686.1 | - | 20 - 21 | 2498 | 103 | ||
| BC009351.1 | - | 23 - 24 | 2995 | 103 | ||
| BC018718.1 | - | 23 - 24 | 2986 | 4123580 | 103 | |
| BC020970.1 | - | 22 - 23 | 2963 | 103 | ||
| X69838.1 | - | 19 - 20 | 2418 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001550.hg.1, TC6_apd_hap1000091.hg.1, TC6_cox_hap2000177.hg.1, TC6_dbb_hap3000166.hg.1, TC6_mcf_hap5000152.hg.1, TC6_qbl_hap6000167.hg.1
Set Membership: |
|

EHMT2
Genomic Map