Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184891.1 | NP_001171820.1 | 3 - 4 | 1073 | 67 | |
| NM_025207.4 | NP_079483.3 | 2 - 3 | 1517 | 67 | ||
| NM_201398.2 | NP_958800.1 | 3 - 4 | 1073 | 67 | ||
| GenBank mRNA | AF481877.1 | - | 2 - 3 | 825 | 67 | |
| AF520568.1 | - | 2 - 3 | 1438 | 67 | ||
| AK090542.1 | - | 2 - 3 | 1459 | 67 | ||
| BC011378.1 | - | 3 - 4 | 1002 | 3855223 | 67 | |
| BC020253.1 | - | 1 - 2 | 352 | 67 | ||
| BC021096.1 | - | 3 - 4 | 991 | 4891331 | 67 | |
| BC032323.1 | - | 3 - 4 | 1008 | 5242727 | 67 | |
| DQ458779.1 | - | 2 - 3 | 1116 | 67 | ||
| U79241.1 | - | 1 - 2 | 668 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001282.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |

FLAD1
Genomic Map