Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC014012.2 | - | 2 - 3 | 895 | 68 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |

FLAD1
Genomic Map