Assay Details
| Gene Symbol: KLHL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198317.2 | NP_938073.1 | 5 - 6 | 931 | 75 | |
| XM_006710600.3 | XP_006710663.1 | 5 - 6 | 1484 | 75 | ||
| XM_006710601.3 | XP_006710664.1 | 5 - 6 | 1484 | 75 | ||
| GenBank mRNA | AK127585.1 | - | 4 - 5 | 578 | 75 | |
| AK293258.1 | - | 1 - 2 | 60 | 75 | ||
| AY423763.1 | - | 5 - 6 | 931 | 75 | ||
| BC105742.1 | - | 2 - 3 | 447 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000021.hg
Additional Information:
This assay is in the 5'UTR region of the AK293258.1,BC105742.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000617073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000175.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000175.hg.1 | ENST00000455747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000175.hg.1 | ENST00000473277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000175.hg.1 | ENST00000540863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000175.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000175.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000178.hg.1 | ENST00000338591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000178.hg.1 | ENST00000455747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000178.hg.1 | ENST00000473277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000178.hg.1 | ENST00000540863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000178.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000178.hg.1 | TC01000021.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144391.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144394.hg.1 | TC0100006479.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000338591.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000463212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000466300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000481067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000617073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | ENST00000622660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006479.hg.1 | NM_198317.2 | Refseq |

KLHL17
Genomic Map