Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182981.2 | NP_892026.1 | 1 - 2 | 147 | 103 | |
| GenBank mRNA | AK290383.1 | - | 1 - 2 | 129 | 103 | |
| AY037158.1 | - | 1 - 2 | 112 | 103 | ||
| AY258068.1 | - | 1 - 2 | 128 | 103 | ||
| BC020434.1 | - | 1 - 2 | 113 | 103 | ||
| BC032476.1 | - | 1 - 2 | 112 | 5215262 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000652.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290383.1,AY037158.1,AY258068.1,BC032476.1,NM_182981.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16009176.hg.1 | ENST00000393306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009176.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009176.hg.1 | TC16000652.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009177.hg.1 | ENST00000343939.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009177.hg.1 | ENST00000361711.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009177.hg.1 | ENST00000393306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009177.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009177.hg.1 | TC16000652.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009175.hg.1 | ENST00000393306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009175.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009175.hg.1 | TC16000652.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009174.hg.1 | ENST00000393306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009174.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009174.hg.1 | TC16000652.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | TC1600011440.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191663.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | TC1600011440.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191668.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | TC1600011440.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600191662.hg.1 | NM_182981.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 193082 | - | - |

OSGIN1
Genomic Map