Assay Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083592.1 | NP_001077061.1 | 2 - 3 | 554 | 88 | |
| NM_005012.3 | NP_005003.2 | 2 - 3 | 554 | 88 | ||
| XM_011541526.1 | XP_011539828.1 | 2 - 3 | 153 | 88 | ||
| GenBank mRNA | AK308088.1 | - | 2 - 3 | 560 | 88 | |
| BC080541.1 | - | 2 - 3 | 554 | 88 | ||
| BC128386.1 | - | 3 - 4 | 164 | 88 | ||
| M97675.1 | - | 2 - 3 | 530 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000723.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011541526.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein tyrosine kinase signaling pathway
peptidyl-tyrosine phosphorylation
Wnt signaling pathway, planar cell polarity pathway
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
Wnt-protein binding
coreceptor activity involved in Wnt signaling pathway, planar cell polarity pathway
transmembrane receptor protein tyrosine kinase signaling pathway
peptidyl-tyrosine phosphorylation
Wnt signaling pathway, planar cell polarity pathway
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
Wnt-protein binding
coreceptor activity involved in Wnt signaling pathway, planar cell polarity pathway
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01011482.hg.1 | ENST00000371079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011482.hg.1 | ENST00000371080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011482.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011482.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011482.hg.1 | TC01000723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011484.hg.1 | ENST00000371079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011484.hg.1 | ENST00000371080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011484.hg.1 | ENST00000544776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011484.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011484.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011484.hg.1 | TC01000723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011480.hg.1 | ENST00000371079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011480.hg.1 | ENST00000371080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011480.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011480.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011480.hg.1 | TC01000723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | ENST00000371079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | ENST00000371080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | ENST00000544776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011486.hg.1 | TC01000723.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | ENST00000371079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | ENST00000371080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | ENST00000544776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011487.hg.1 | TC01000723.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008594.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160978.hg.1 | TC0100008594.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160982.hg.1 | TC0100008594.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160979.hg.1 | TC0100008594.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000371079.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000371080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000482426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | ENST00000545203.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | NM_001083592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | NM_005012.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160983.hg.1 | TC0100008594.hg.1 | Clariom D TC |

ROR1
Genomic Map