| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
ENST00000265735.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
ENST00000415788.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
ENST00000428696.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
ENST00000437425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
ENST00000445866.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
ENST00000447873.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
NM_001099400.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
NM_001099401.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
NM_003919.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024485.hg.1 |
TC07001614.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000265735.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000415788.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000428696.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000437425.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000445866.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000447873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000472326.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_001099400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_001099401.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_001301139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_003919.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000265735.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000415788.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000428696.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000437425.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000445866.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000447873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
ENST00000472326.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_001099400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_001099401.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_001301139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
NM_003919.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184481.hg.1 |
TC0700011821.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000265735.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000415788.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000428696.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000437425.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000445866.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000447873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000462731.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000522045.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_001099400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_001099401.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_001301139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_003919.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000265735.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000415788.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000428696.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000437425.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000445866.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000447873.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000462731.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
ENST00000522045.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_001099400.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_001099401.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_001301139.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
NM_003919.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184485.hg.1 |
TC0700011821.hg.1 |
Clariom D TC |