Assay Details
| Gene Symbol: SORBS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716266.1 | XP_006716329.1 | 1 - 2 | 146 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006716266.1 transcripts.
Set Membership: |
|

SORBS3
Genomic Map