Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135703.2 | NP_001129175.1 | 3 - 4 | 544 | 111 | |
| NM_013437.4 | NP_038465.1 | 4 - 5 | 601 | 111 | ||
| GenBank mRNA | AB208957.1 | - | 2 - 3 | 276 | 111 | |
| AF064841.1 | - | 3 - 4 | 539 | 111 | ||
| AF166350.1 | - | 4 - 5 | 516 | 111 | ||
| AK289752.1 | - | 4 - 5 | 505 | 111 | ||
| AK299374.1 | - | 3 - 4 | 541 | 111 | ||
| BC032109.2 | - | 4 - 5 | 477 | 4842797 | 111 | |
| DC372924.1 | - | 3 - 4 | 541 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001511.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019867.hg.1 | ENST00000276654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019867.hg.1 | ENST00000424843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019867.hg.1 | ENST00000520873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019867.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019867.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019867.hg.1 | TC08001511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019866.hg.1 | ENST00000276654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019866.hg.1 | ENST00000424843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019866.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019866.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019866.hg.1 | TC08001511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019868.hg.1 | ENST00000276654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019868.hg.1 | ENST00000424843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019868.hg.1 | ENST00000520873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019868.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019868.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019868.hg.1 | TC08001511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019864.hg.1 | ENST00000276654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019864.hg.1 | ENST00000424843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019864.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019864.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019864.hg.1 | TC08001511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019865.hg.1 | ENST00000276654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019865.hg.1 | ENST00000424843.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019865.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019865.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08019865.hg.1 | TC08001511.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177702.hg.1 | TC0800011395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177703.hg.1 | TC0800011395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800177704.hg.1 | TC0800011395.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |

LRP12
Genomic Map