Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182614.2 | NP_872420.1 | 2 - 3 | 217 | 94 | |
| XM_005272413.3 | XP_005272470.1 | 2 - 3 | 243 | 94 | ||
| XM_005272415.2 | XP_005272472.1 | 2 - 3 | 217 | 94 | ||
| XM_017020558.1 | XP_016876047.1 | 2 - 3 | 243 | 94 | ||
| GenBank mRNA | BC018995.2 | - | 2 - 3 | 217 | 4300679 | 94 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005272415.2 transcripts.
Set Membership: |
|

TMEM255B
Genomic Map