Assay Details
| Gene Symbol: BHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178005.1 | NP_001171476.1 | 4 - 5 | 462 | 60 | |
| NM_017614.4 | NP_060084.2 | 5 - 6 | 654 | 60 | ||
| GenBank mRNA | AF257473.1 | - | 5 - 6 | 629 | 60 | |
| AK000008.1 | - | 4 - 5 | 566 | 60 | ||
| AK298298.1 | - | 4 - 5 | 419 | 60 | ||
| AK315520.1 | - | 5 - 6 | 612 | 60 | ||
| BC020665.1 | - | 5 - 6 | 609 | 4722608 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000386.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sulfur amino acid metabolic process
amino-acid betaine metabolic process
methylation
S-methylmethionine metabolic process
S-adenosylmethionine metabolic process
L-methionine salvage
zinc ion binding
betaine-homocysteine S-methyltransferase activity
S-methylmethionine-homocysteine S-methyltransferase activity
sulfur amino acid metabolic process
amino-acid betaine metabolic process
methylation
S-methylmethionine metabolic process
S-adenosylmethionine metabolic process
L-methionine salvage
zinc ion binding
betaine-homocysteine S-methyltransferase activity
S-methylmethionine-homocysteine S-methyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005433.hg.1 | ENST00000255192.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005433.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005433.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005433.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005433.hg.1 | TC05000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005434.hg.1 | ENST00000255192.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005434.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005434.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005434.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005434.hg.1 | TC05000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005430.hg.1 | ENST00000255192.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005430.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005430.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005430.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05005430.hg.1 | TC05000386.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000518758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | ENST00000523472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007875.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153897.hg.1 | TC0500007875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000523046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000255192.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000519743.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000521567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | ENST00000523046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | NM_001178005.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | NM_017614.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500153894.hg.1 | TC0500007875.hg.1 | Clariom D TC |

BHMT2
Genomic Map