Assay Details
| Gene Symbol: BFSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161705.1 | NP_001155177.1 | 4 - 5 | 522 | 142 | |
| NM_001195.4 | NP_001186.1 | 4 - 5 | 666 | 142 | ||
| NM_001278606.1 | NP_001265535.1 | 5 - 6 | 657 | 142 | ||
| NM_001278607.1 | NP_001265536.1 | 4 - 5 | 518 | 142 | ||
| NM_001278608.1 | NP_001265537.1 | 6 - 7 | 643 | 142 | ||
| GenBank mRNA | AF039655.1 | - | 4 - 5 | 626 | 142 | |
| AK304673.1 | - | 6 - 7 | 643 | 142 | ||
| AK316247.1 | - | 5 - 6 | 535 | 142 | ||
| BC041483.1 | - | 4 - 5 | 522 | 5406467 | 142 | |
| BQ230198.1 | - | 4 - 5 | 361 | 142 | ||
| Y16717.2 | - | 4 - 5 | 626 | 142 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000655.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | ENST00000377868.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | ENST00000377873.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | ENST00000536626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | ENST00000544874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20009177.hg.1 | TC20000655.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000473415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008493.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001278608.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | TC2000008493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000377868.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000377873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000492424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | ENST00000536626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001161705.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001195.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001278606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001278607.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000159604.hg.1 | NM_001278608.1 | Refseq |

BFSP1
Genomic Map