Assay Details
| Gene Symbol: SIRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080816.2 | NP_543006.2 | 2 - 3 | 112 | ||
| GenBank mRNA | BC020629.1 | - | 2 - 3 | 477 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000555.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | ENST00000303415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | ENST00000344103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | ENST00000381580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | ENST00000381583.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008059.hg.1 | TC20000555.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | ENST00000303415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | ENST00000344103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | ENST00000381583.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008062.hg.1 | TC20000555.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | ENST00000303415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | ENST00000344103.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | ENST00000381583.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008061.hg.1 | TC20000555.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | TC2000009971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170931.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | TC2000009971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170928.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | TC2000009971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000170930.hg.1 | NM_080816.2 | Refseq |

SIRPG
Genomic Map