| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010679.hg.1 |
ENST00000409663.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010679.hg.1 |
ENST00000409959.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010679.hg.1 |
ENST00000440693.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010679.hg.1 |
NM_001131007.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010679.hg.1 |
NM_015196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010679.hg.1 |
TC04000774.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000240487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000409663.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000409959.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000478919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
NM_001131007.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
NM_015196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000240487.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000409663.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000409959.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
ENST00000478919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
NM_001131007.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
NM_015196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400162072.hg.1 |
TC0400009038.hg.1 |
Clariom D TC |