Assay Details
| Gene Symbol: ADGRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270666.4 | XP_005270723.1 | 9 - 10 | 2751 | 90 | |
| XM_006710485.3 | XP_006710548.1 | 10 - 11 | 2767 | 90 | ||
| XM_006710488.2 | XP_006710551.1 | 10 - 11 | 2061 | 90 | ||
| XM_006710489.3 | XP_006710552.1 | 10 - 11 | 1952 | 90 | ||
| XM_017000782.1 | XP_016856271.1 | 14 - 15 | 2635 | 90 | ||
| XM_017000783.1 | XP_016856272.1 | 11 - 12 | 2046 | 90 | ||
| XM_017000784.1 | XP_016856273.1 | 10 - 11 | 1951 | 90 | ||
| XM_017000785.1 | XP_016856274.1 | 10 - 11 | 2058 | 90 | ||
| XM_017000786.1 | XP_016856275.1 | 10 - 11 | 2062 | 90 | ||
| XM_017000787.1 | XP_016856276.1 | 10 - 11 | 1973 | 90 | ||
| XM_017000790.1 | XP_016856279.1 | 11 - 12 | 1987 | 90 | ||
| XM_017000791.1 | XP_016856280.1 | 9 - 10 | 2051 | 90 | ||
| XM_017000793.1 | XP_016856282.1 | 11 - 12 | 1989 | 90 | ||
| XM_017000794.1 | XP_016856283.1 | 11 - 12 | 1988 | 90 | ||
| XM_017000795.1 | XP_016856284.1 | 11 - 12 | 1987 | 90 | ||
| XM_017000796.1 | XP_016856285.1 | 9 - 10 | 2011 | 90 | ||
| GenBank mRNA | AB018329.1 | - | 4 - 5 | 502 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ADGRL2
Genomic Map