Assay Details
| Gene Symbol: ERMAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017922.1 | NP_001017922.1 | 8 - 9 | 907 | 95 | |
| NM_018538.3 | NP_061008.2 | 7 - 8 | 803 | 95 | ||
| XM_006710313.3 | XP_006710376.1 | 9 - 10 | 1172 | 95 | ||
| XM_011540570.2 | XP_011538872.1 | 8 - 9 | 863 | 95 | ||
| GenBank mRNA | AF311285.1 | - | 7 - 8 | 803 | 95 | |
| AK056138.1 | - | 8 - 9 | 881 | 95 | ||
| AK314436.1 | - | 8 - 9 | 891 | 95 | ||
| AY049028.1 | - | 7 - 8 | 821 | 95 | ||
| BC099703.1 | - | 7 - 8 | 736 | 40000626 | 95 | |
| BC099707.1 | - | 7 - 8 | 741 | 40000628 | 95 | |
| BC099712.1 | - | 7 - 8 | 736 | 40000629 | 95 | |
| BC099713.1 | - | 7 - 8 | 736 | 40000630 | 95 | |
| BX537371.1 | - | 5 - 6 | 1405 | 95 | ||
More Information
Set Membership: |
|

ERMAP
Genomic Map