| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003420.hg.1 |
ENST00000377067.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003420.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003420.hg.1 |
TC13000323.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
ENST00000618283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
ENST00000618283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152060.hg.1 |
TC1300007707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000483422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
ENST00000618596.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007707.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152059.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152059.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152059.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152059.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152059.hg.1 |
TC1300007707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300152062.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152062.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152062.hg.1 |
ENST00000377067.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152062.hg.1 |
NM_004466.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152062.hg.1 |
TC1300007707.hg.1 |
Clariom D TC |