| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023143.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023143.hg.1 |
ENST00000513319.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023143.hg.1 |
ENST00000543620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023143.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023143.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05023143.hg.1 |
TC05001714.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011852.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000231004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000503759.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000505593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000508067.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
ENST00000513319.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
NM_001178102.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
NM_002317.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181729.hg.1 |
TC0500011852.hg.1 |
Clariom D TC |